00500590 Fluka

Chlorogenic acid

primary reference standard

DOWNLOAD MSDS (PDF)

Synonym: 1,3,4,5-Tetrahydroxycyclohexanecarboxylic acid 3-(3,4-dihydroxycinnamate), 3-(3,4-Dihydroxycinnamoyl)quinic acid

  • CAS Number 327-97-9

  • Empirical Formula (Hill Notation) C16H18O9

  • Molecular Weight 354.31

  •  EC Number 206-325-6

  •  MDL number MFCD00003862

Price and Availability

Customers Also Viewed

Caffeic acid

≥98.0% (HPLC)

Chlorogenic acid

≥95% (titration)

Neochlorogenic acid

≥98.0% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?