EMAIL THIS PAGE TO A FRIEND

00740580 Fluka

Quercitrin

primary pharmaceutical reference standard

Synonym: Quercetin 3-rhamnoside

  • CAS Number 522-12-3

  • Empirical Formula (Hill Notation) C21H20O11

  • Molecular Weight 448.38

  •  Beilstein Registry Number 68135

  •  EC Number 208-322-5

  •  MDL number MFCD00016932

Purchase

Description

Application

Reference standard in the analysis of herbal medicinal products.

General description

Produced and qualified by HWI Analytik
Exact content by quantitative NMR can be found on the certificate

Price and Availability

Customers Also Viewed

Hyperoside
00180585Hyperoside

primary pharmaceutical reference standard

Isoquercitrin

primary pharmaceutical reference standard

Quercetin 3-<SC>D</SC>-galactoside

≥97.0% (HPLC)

Quercetin 3-glucoside

analytical standard

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
UZ5950000

Documents

Certificate of Analysis

Protocols & Articles

Articles

Reference Standards for Herbal Medicinal Products

In collaborationwith HWI ANALYTIK, Sigma-Aldrich is launching a new product line of primary reference standards for use in the quality control, in-process control and stability testing of herbalmedic...
Matthias Nold
AnalytiX Volume 10 Article 1
Keywords: High performance liquid chromatography, Karl Fischer titrations, Nuclear magnetic resonance spectroscopy, PAGE, Pharmaceutical, Solvents, Titrations

Peer-Reviewed Papers

References

Set your institution to view full text papers.

[Chemical constituents from Commelina communis]. Yuan HE, Zhou XD, Meng LJ, et al. Zhongguo Zhong Yao Za Zhi 38(19), 3304-8, (2013)

Loading...


[Chemical constituents of Rhododendron seniavinii]. Wang QQ, Zhang Y, Ye WC, et al. Zhongguo Zhong Yao Za Zhi 38(3), 366-70, (2013)

Loading...


[Thermosensitization of tumor cells with inhibitors of chaperone activity and expression]. Kudriavtsev VA, Makarova IuM, and Kabakav AE Biomed. Khim. 58(6), 662-72, (2012)

Loading...


[Tracking the active component of Tebatan medicine Meconopsis quintuplinervia from Gansu]. Cheng F, Guo M, Zhang Y, et al. Zhong Yao Cai 34(1), 69-71, (2011)

Loading...


[Studies on the flavonoidls from the herb of Striga asiatica]. Huang S, Chen JH, Gong M, et al. Zhong Yao Cai 33(7), 1089-91, (2010)

Loading...


[Studies on the chemical constituents of Lithocarpus polystachyus]. Li SH, Wu XJ, Yang QD, et al. Zhong Yao Cai 33(4), 549-51, (2010)

Loading...


Lessons learned from herbal medicinal products: the example of St. John's Wort (perpendicular). Adolf Nahrstedt et al J. Nat. Prod. 73, 1015-21, (2010)

Loading...


Quercitrin protects skin from UVB-induced oxidative damage. Yin Y, Li W, Son YO, et al. Toxicol. Appl. Pharmacol. 269(2), 89-99, (2013)

Loading...


The leishmanicidal flavonols quercetin and quercitrin target Leishmania (Leishmania) amazonensis arginase. da Silva ER, Maquiaveli Cdo C, and Magalhães PP Exp. Parasitol. 130(3), 183-8, (2012)

Loading...


Inhibition of 3(17)alpha-hydroxysteroid dehydrogenase (AKR1C21) by aldose reductase inhibitors. Urmi Dhagat et al Bioorg. Med. Chem. 16, 3245-54, (2008)

Loading...


Nanoencapsulation and characterization of Albizia chinensis isolated antioxidant quercitrin on PLA nanoparticles. Kumari A, Yadav SK, Pakade YB, et al. Colloids Surf. B Biointerfaces 82(1), 224-32, (2011)

Loading...


Kalanchoe pinnata inhibits mast cell activation and prevents allergic airway disease. Cruz EA, Reuter S, Martin H, et al. Phytomedicine 19(2), 115-21, (2012)

Loading...


Erigeroflavanone, a flavanone derivative from the flowers of Erigeron annuus with protein glycation and aldose reductase inhibitory activity. Nam Hee Yoo et al J. Nat. Prod. 71, 713-5, (2008)

Loading...


Reliability of bond dissociation enthalpy calculated by the PM6 method and experimental TEAC values in antiradical QSAR of flavonoids. Dragan Ami? et al Bioorg. Med. Chem. 18, 28-35, (2010)

Loading...


An efficient and economical MTT assay for determining the antioxidant activity of plant natural product extracts and pure compounds. Yunbao Liu et al J. Nat. Prod. 73, 1193-5, (2010)

Loading...


Protective effect of olive leaf extract on hippocampal injury induced by transient global cerebral ischemia and reperfusion in Mongolian gerbils. Dekanski D, Selaković V, Piperski V, et al. Phytomedicine 18(13), 1137-43, (2011)

Loading...


Two new hepatoprotective stilbene glycosides from Acer mono leaves. Hyekyung Yang et al J. Nat. Prod. 68, 101-3, (2005)

Loading...


Inhibitors of osteoclast differentiation from Cephalotaxus koreana. Kee Dong Yoon et al J. Nat. Prod. 70, 2029-32, (2007)

Loading...


Sweeping under controlled electroosmotic flow and micellar electrokinetic chromatography for on-line concentration and determination of trace phlorizin and quercitrin in urine samples. Zhang X and Zhang Z J. Pharm. Biomed. Anal. 56(2), 330-5, (2011)

Loading...


Eucalmaidins A-E, (+)-oleuropeic acid derivatives from the fresh leaves of Eucalyptus maideni. Li-Wen Tian et al J. Nat. Prod. 72, 1608-11, (2009)

Loading...


HIV-1 ribonuclease H inhibitory phenolic glycosides from Eugenia hyemalis. Heidi R Bokesch et al J. Nat. Prod. 71, 1634-6, (2008)

Loading...


Studies on the chemical constituents and anticancer activity of Saxifraga stolonifera (L) Meeb. Zhuo Chen et al Bioorg. Med. Chem. 16, 1337-44, (2008)

Loading...


Chemical constituents of Indonesian plant Protium javanicum Burm. f. and their antifeedant activities against Coptotermes formosanus Shiraki. Adfa M, Hattori Y, Ninomiya M, et al. Nat. Prod. Res. 27(3), 270-3, (2013)

Loading...


Quercitrin protects against ultraviolet B-induced cell death in vitro and in an in vivo zebrafish model. Yang HM, Ham YM, Yoon WJ, et al. J. Photochem. Photobiol. B, Biol. 114, 126-31, (2012)

Loading...


New Hedgehog/GLI signaling inhibitors from Excoecaria agallocha. Yusnita Rifai et al Bioorg. Med. Chem. Lett. 21, 718-22, (2011)

Loading...


Theoretical investigation of the effect of sugar substitution on the antioxidant properties of flavonoids. Lespade L and Bercion S Free Radic. Res. 46(3), 346-58, (2012)

Loading...


Campomanesia lineatifolia Ruiz & Pav. as a gastroprotective agent. Madalosso RC, Oliveira GC, Martins MT, et al. J. Ethnopharmacol. 139(3), 772-9, (2012)

Loading...


Quercetin and quercitrin protect against cytokine‑induced injuries in RINm5F β-cells via the mitochondrial pathway and NF-κB signaling. Dai X, Ding Y, Zhang Z, et al. Int. J. Mol. Med. 31(1), 265-71, (2013)

Loading...


Quercitrin and taxifolin stimulate osteoblast differentiation in MC3T3-E1 cells and inhibit osteoclastogenesis in RAW 264.7 cells. Satué M, Arriero Mdel M, Monjo M, et al. Biochem. Pharmacol. 86(10), 1476-86, (2013)

Loading...


Pim2 inhibitors from the Papua New Guinean plant Cupaniopsis macropetala. Rohan A Davis et al J. Nat. Prod. 71, 451-2, (2008)

Loading...


Stimulation of activity and expression of antioxidant enzymes by solvent fractions and isolated compound from Cedrela sinensis leaves in HepG2 cells. Bak MJ, Jeong JH, Kang HS, et al. J. Med. Food 14(4), 405-12, (2011)

Loading...


Evaluation of flavonoid contents and antioxidant capacity of the aerial parts of common and tartary buckwheat plants. Zielińska D, Turemko M, Kwiatkowski J, et al. Molecules 17(8), 9668-82, (2012)

Loading...


Simultaneous extraction and separation of flavonols and flavones from Chamaecyparis obtusa by multi-phase extraction using an ionic liquid-modified microsphere polymer. Tian M, Bi W, and Row KH Phytochem. Anal. 23(6), 576-81, (2012)

Loading...


Identification of quercitrin as an inhibitor of the p90 S6 ribosomal kinase (RSK): structure of its complex with the N-terminal domain of RSK2 at 1.8 Å resolution. Derewenda U, Artamonov M, Szukalska G, et al. Acta Crystallogr. D Biol. Crystallogr. 69(Pt 2), 266-75, (2013)

Loading...


Quercitrin a bioflavonoid improves the antioxidant status in streptozotocin: induced diabetic rat tissues. Babujanarthanam R, Kavitha P, Mahadeva Rao US, et al. Mol. Cell Biochem. 358(1-2), 121-9, (2011)

Loading...


HPLC analysis of kaempherol and quercetin derivatives isolated by different extraction techniques from plant matrix. Skalicka-Woźniak K, Szypowski J, and Głowniak K J. AOAC Int. 94(1), 17-21, (2011)

Loading...


Separation of the antioxidant compound quercitrin from Lindera obtusiloba Blume and its antimelanogenic effect on B16F10 melanoma cells. Hong CO, Lee HA, Rhee CH, et al. Biosci. Biotechnol. Biochem. 77(1), 58-64, (2013)

Loading...


Protective effect of components isolated from Lindera erythrocarpa against oxidative stress-induced apoptosis of H9c2 cardiomyocytes. Kim JA, Jung YS, Kim MY, et al. Phytother Res. 25(11), 1612-7, (2011)

Loading...


Construction of an Indonesian herbal constituents database and its use in Random Forest modelling in a search for inhibitors of aldose reductase. Sadaf Naeem et al Bioorg. Med. Chem. 20, 1251-8, (2012)

Loading...


Suppression of nitric oxide production on LPS/IFN-?-stimulated RAW264.7 macrophages by a novel catechin, pilosanol N, from Agrimonia pilosa Ledeb. Junsei Taira et al Bioorg. Med. Chem. Lett. 22, 1766-9, (2012)

Loading...


A novel flavonoid from Lespedeza virgata (Thunb.) DC.: structural elucidation and antioxidative activity. Li Tan et al Bioorg. Med. Chem. Lett. 17, 6311-5, (2007)

Loading...


Alkaloids and flavonoids of Argemone mexicana. Singh S, Pandey VB, and Singh TD Nat. Prod. Res. 26(1), 16-21, (2012)

Loading...


Merck 14,8037

Beil. 18,V,5,514

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?