00797 Fluka

Methyl-trioctylammonium bis(trifluoromethylsulfonyl)imide

for electrochemistry, ≥99% (NMR)

DOWNLOAD MSDS (PDF)

Synonym: Methyltri-n-octylammonium bis(trifluoromethanesulfonyl)imide, Trioctylmethylammonium bis(trifluoromethylsulfonyl)imide

  • CAS Number 375395-33-8

  • Empirical Formula (Hill Notation) C27H54F6N2O4S2

  • Molecular Weight 648.85

  •  MDL number MFCD08275364

  •  PubChem Substance ID 24845076

Properties

Related Categories Ammonium, Chemical Synthesis, Ionic Liquids, Specialty Synthesis
grade   for electrochemistry
assay   ≥99% (NMR)
electrochemical window   -2.6-+2.8 V
impurities   ≤300 ppm water
refractive index   n20/D 1.439
conductivity   ~0.04 mS/cm
anion traces   bromide (Br-): ≤20 mg/kg
  chloride (Cl-): ≤20 mg/kg
  fluoride (F-): ≤20 mg/kg
  nitrate (NO3-): ≤10 mg/kg
  sulfate (SO42-): ≤10 mg/kg

Price and Availability

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?