CAS Number 928-91-6
Empirical Formula (Hill Notation) C6H12O
Molecular Weight 100.16
Beilstein Registry Number 1720136
EC Number 213-187-0
MDL number MFCD00148974
PubChem Substance ID 24845108
| Related Categories | Acyclic, Alkenes, Building Blocks, Chemical Synthesis, Organic Building Blocks More... |
| assay | ≥95.0% (GC) |
| refractive index |
n |
| bp | 80 °C/30 mmHg(lit.) |
| density | 0.857 g/mL at 20 °C(lit.) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?