Synonym: 2-Butynedioic acid
CAS Number 142-45-0
Linear Formula HOOCC≡CCOOH
Molecular Weight 114.06
Beilstein Registry Number 878357
EC Number 205-536-0
MDL number MFCD00004362
Popular Documents: RAMAN FTIR (PDF)
| Related Categories | Building Blocks, C1 to C5, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis, |
| grade | purum |
| assay | ≥94.0% (T) |
| mp | 180-187 °C (dec.)(lit.) |
| storage temp. | 2-8°C |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?