Synonym: Bergamotine, Bergaptin
CAS Number 7380-40-7
Empirical Formula (Hill Notation) C21H22O4
Molecular Weight 338.40
Beilstein Registry Number 1353914
MDL number MFCD00017409
PubChem Substance ID 24845158
| Related Categories | A - D, Alphabetical, Analytical Standards, Analytical/Chromatography, B, |
| grade | analytical standard, for terpene analysis |
| assay | ≥98.0% (HPLC) |
| mp | 75-80 °C |
| storage temp. | 2-8°C |
| Gene Information | human ... CYP3A4(1576) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?