Synonym:
(S)
CAS Number 23513-08-8
Empirical Formula (Hill Notation) C19H30O4
Molecular Weight 322.44
Beilstein Registry Number 2059802
MDL number MFCD00877794
| Related Categories | Aldehydes and Ketones, Alphabetical, Analytical Standards, Analytical/Chromatography, Chromatography, |
| grade | analytical standard |
| assay | ≥95.0% (HPLC) |
| shelf life | (limited shelf life, expiry date on the label) |
| storage temp. | 2-8°C |
[8]-Gingerol is a bioactive compound found in ginger (Zingiber officinale) with anti-inflammatory and antioxidant activity.1
Bottomless glass bottle. Contents are inside inserted fused cone.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?