02390 Fluka

1,2-Ethanedithiol

≥98.0% (GC)

DOWNLOAD MSDS (PDF)

Synonym: 1,2-Dimercaptoethane, Dithioglycol, Ethylene mercaptan

  • CAS Number 540-63-6

  • Linear Formula HSCH2CH2SH

  • Molecular Weight 94.20

  •  Beilstein Registry Number 505946

  •  EC Number 208-752-3

  •  MDL number MFCD00004892

  •  PubChem Substance ID 24845272

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds, Thiols/Mercaptans More...
vapor density   >1 (vs air)
vapor pressure   4.8 mmHg ( 20 °C)
assay   ≥98.0% (GC)
refractive index   n20/D 1.558(lit.)
  n20/D 1.558
bp   144-146 °C(lit.)
mp   −41 °C(lit.)
density   1.123 g/mL at 25 °C(lit.)

Price and Availability

Customers Also Viewed

1,2-Ethanedithiol

technical grade, ≥90%

Thioanisole

purum, ≥99.0% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?