02473 Fluka

2-Hexanone

analytical standard

DOWNLOAD MSDS (PDF)

Synonym: Butyl methyl ketone

  • CAS Number 591-78-6

  • Linear Formula CH3(CH2)3COCH3

  • Molecular Weight 100.16

  •  Beilstein Registry Number 1737676

  •  EC Number 209-731-1

  •  MDL number MFCD00009482

  •  PubChem Substance ID 24845297

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Alpha Sort, Analytical Standards, Analytical/Chromatography, Chromatography, Cosmetic Standards,
grade   analytical standard
vapor pressure   10 mmHg ( 39 °C)
assay   ≥99.5% (GC)
shelf life   (limited shelf life, expiry date on the label)
refractive index   n20/D 1.401(lit.)
  n20/D 1.401
bp   127 °C(lit.)
mp   −57 °C(lit.)
density   0.812 g/mL at 25 °C(lit.)

Price and Availability

Proficiency Testing Samples from RTC


PT Samples from RTCRTC has been producing environmental LPTP for more than 20 years and send out over 20,000 PT samples a year, throughout
the world.





Customers Also Viewed

2-Hexanone

purum, ≥96.0% (GC)

2-Hexanone

reagent grade, 98%

200 μg/mL in methanol, analytical standard

4-Methyl-2-pentanone

CHROMASOLV®, for HPLC, ≥99.5%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?