02476 Fluka

2-Heptanone

analytical standard

DOWNLOAD MSDS (PDF)

Synonym: Methyl pentyl ketone

  • CAS Number 110-43-0

  • Linear Formula CH3(CH2)4COCH3

  • Molecular Weight 114.19

  •  Beilstein Registry Number 1699063

  •  EC Number 203-767-1

  •  MDL number MFCD00009513

  •  PubChem Substance ID 24845299

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Alcoholic & Nonalcoholic Beverages, All Alphabetically Sorted, Alpha Sort, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
vapor density   3.94 (vs air)
vapor pressure   2.14 mmHg ( 20 °C)
assay   ≥99.8% (GC)
autoignition temp.   739 °F
shelf life   (limited shelf life, expiry date on the label)
expl. lim.   1.11 %, 65 °F
  7.9 %, 121 °F
refractive index   n20/D 1.408(lit.)
  n20/D 1.409
bp   149-150 °C(lit.)
mp   −35 °C(lit.)
density   0.82 g/mL at 25 °C(lit.)

Price and Availability

Proficiency Testing Samples from RTC


PT Samples from RTCRTC has been producing environmental LPTP for more than 20 years and send out over 20,000 PT samples a year, throughout
the world.





Customers Also Viewed

2-Heptanone

≥98%, FCC, FG

2-Heptanone

99%

3-Heptanone

98%

4-Heptanone

analytical standard, for environmental analysis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?