02487 Fluka

Chloroform

analytical standard

DOWNLOAD MSDS (PDF)

Synonym: Methylidyne trichloride, Trichloromethane

  • CAS Number 67-66-3

  • Empirical Formula (Hill Notation) CHCl3

  • Molecular Weight 119.38

  •  Beilstein Registry Number 1731042

  •  EC Number 200-663-8

  •  MDL number MFCD00000826

  •  PubChem Substance ID 24845306

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Allergens, Alpha Sort, Analytical Standards, Analytical/Chromatography, Applications,
grade   analytical standard
vapor density   4.1 (vs air)
vapor pressure   160 mmHg ( 20 °C)
assay   ≥99.9% (GC)
shelf life   (limited shelf life, expiry date on the label)
contains   ~0.003% amylene as stabilizer
refractive index   n20/D 1.445(lit.)
  n20/D 1.446
bp   60.5-61.5 °C(lit.)
mp   −63 °C(lit.)
density   1.492 g/mL at 25 °C(lit.)

Price and Availability

Proficiency Testing Samples from RTC


PT Samples from RTCRTC has been producing environmental LPTP for more than 20 years and send out over 20,000 PT samples a year, throughout
the world.





Customers Also Viewed

Chloroform

contains 100-200 ppm amylenes as stabilizer, ≥99.5%

Chloroform

CHROMASOLV® Plus, for HPLC, ≥99.9%, contains amylenes as stabilizer

Chloroform

contains ~1% ethanol as stabilizer, for residue analysis, ≥99.8%

Chloroform

puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)

Chloroform

purum, ≥99.5% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?