• USA Home
  • 02559 - Bromocresol Green solution

EMAIL THIS PAGE TO A FRIEND
02559 Fluka

Bromocresol Green solution

indicator, ready-to-use

  • CAS Number 76-60-8

  • Empirical Formula (Hill Notation) C21H14Br4O5S

  • Molecular Weight 698.01

  •  Beilstein Registry Number 372527

  •  EC Number 200-661-7

  •  MDL number MFCD00005874

  •  PubChem Substance ID 24845317

  • Popular Documents:  FTIR (PDF)  

Purchase

Description

Application

ready-to-use spray and dip reagent for chromatography

Legal Information

FLUKA is a registered trademark of Honeywell Specialty Chemicals Seelze GmbH.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1219 - class 3 - PG 2 - Isopropanol, solution
WGK Germany 
2
Flash Point(F) 
53.6 °F
Flash Point(C) 
12 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?