CAS Number 76-54-0
Empirical Formula (Hill Notation) C20H10Cl2O5
Molecular Weight 401.20
Beilstein Registry Number 58009
MDL number MFCD00005047
PubChem Substance ID 24845334
Popular Documents: FTIR (PDF)
| Related Categories | Alphabetical, Analytical Reagents, Analytical/Chromatography, Biochemicals and Reagents, Derivatization Reagents, |
| grade | indicator |
| density | 0.79 g/mL at 20 °C |
ready-to-use spray and dip reagent for chromatography
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?