Synonym: Ferric chloride solution, Iron(III) chloride solution
CAS Number 7705-08-0
Linear Formula FeCl3
Molecular Weight 162.20
MDL number MFCD00011005
PubChem Substance ID 24845365
| Related Categories | 1.00 g/L, AAS, AAS Concentrates, Analytical Standards, Analytical/Chromatography, |
| quality | ampule |
| shelf life | (limited shelf life, expiry date on the label) |
| concentration | 1.00 g/L |
| solubility | H2O: soluble |
prepared with FeCl3.6H2O, HCl and H2O
Dilute the ampule to approx. 850 mL with water. Carefully add 100 mL conc. HCl to acidify. Bring to volume with water.
Customers Also Viewed
TraceCERT®, 1000 mg/L Fe in nitric acid
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?