02799 Fluka

Tetramethylammonium sulfate

for ion pair chromatography, ≥99.0% (T)

DOWNLOAD MSDS (PDF)

Synonym: Bis(tetramethylammonium) sulfate

  • CAS Number 14190-16-0

  • Linear Formula [(CH3)4N]2(SO4)

  • Molecular Weight 244.35

  •  Beilstein Registry Number 3918039

  •  EC Number 238-043-4

  •  MDL number MFCD00012139

  •  PubChem Substance ID 24845386

Properties

Related Categories Acidic Ion Pair Reagents (Anionic), Analytical Reagents, Analytical/Chromatography, Buffers & Reagents, Chromatography Reagents & Solvents,
grade   for ion pair chromatography
assay   ≥99.0% (T)
λ   10 % in H2O
UV absorption   λ: 210 nm Amax: ≤0.05
  λ: 220 nm Amax: ≤0.04
  λ: 230 nm Amax: ≤0.03
  λ: 260 nm Amax: ≤0.02
  λ: 500 nm Amax: ≤0.02
suitability   corresponds to standard for RP gradient test
  corresponds to standard for filter test

Price and Availability

Customers Also Viewed

Tetramethylammonium bisulfate

for ion pair chromatography, ≥99.0% (T)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?