03128 Fluka

Diethylene glycol

analytical standard

DOWNLOAD MSDS (PDF)

Synonym: 2,2′-Oxydiethanol, 2-Hydroxyethyl ether, Bis(2-hydroxyethyl) ether, Diglycol

  • CAS Number 111-46-6

  • Linear Formula (HOCH2CH2)2O

  • Molecular Weight 106.12

  •  Beilstein Registry Number 969209

  •  EC Number 203-872-2

  •  MDL number MFCD00002882

  •  PubChem Substance ID 24845427

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Alpha Sort, Analytical Standards, Analytical/Chromatography, Chromatography, Cosmetic Standards,
grade   analytical standard
vapor density   2.14 (vs air)
vapor pressure   0.01 mmHg ( 20 °C)
description   non-ionic
assay   ≥99.5% (GC)
autoignition temp.   442 °F
shelf life   (limited shelf life, expiry date on the label)
expl. lim.   2-12.3 %
refractive index   n20/D 1.447(lit.)
  n20/D 1.447
bp   245 °C(lit.)
mp   −10 °C(lit.)
density   1.118 g/mL at 25 °C(lit.)

Price and Availability

Proficiency Testing Samples from RTC


PT Samples from RTCRTC has been producing environmental LPTP for more than 20 years and send out over 20,000 PT samples a year, throughout
the world.





Customers Also Viewed

Diethylene glycol

BioUltra, ≥99.0% (GC)

Diethylene glycol

puriss. p.a., ≥99.0% (GC), colorless

Diethylene glycol

ReagentPlus®, 99%

Ethylene glycol

anhydrous, 99.8%

Ethylene glycol

analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?