03440 Fluka

N-Ethyldiisopropylamine

BASF quality, ≥98.0%

DOWNLOAD MSDS (PDF)

Synonym: N,N-Diisopropylethylamine, Hünigs base

  • CAS Number 7087-68-5

  • Linear Formula CH3CH2N[CH(CH3)2]2

  • Molecular Weight 129.24

  •  Beilstein Registry Number 605301

  •  EC Number 230-392-0

  •  MDL number MFCD00008868

  •  PubChem Substance ID 24845480

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Chemical Synthesis, Organic Bases, Synthetic Reagents More...
grade   BASF quality
assay   ≥98.0%
impurities   ≤0.5% water
refractive index   n20/D 1.414
bp   126-128 °C(lit.)
density   0.755 g/mL at 20 °C(lit.)
  0.755 g/mL at 20 °C
  0.757 g/mL at 25 °C(lit.)

Price and Availability

Customers Also Viewed

<I>N</I>,<I>N</I>-<WBR>Diisopropylethylamine

purified by redistillation, 99.5%

<I>N</I>,<I>N</I>-<WBR>Diisopropylethylamine

99.5%, biotech. grade

<I>N</I>,<I>N</I>-<WBR>Diisopropylethylamine

ReagentPlus®, 99%

<I>N</I>-<WBR>Ethyldiisopropylamine

produced by BASF, ≥98.0% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?