EMAIL THIS PAGE TO A FRIEND

03550 Fluka

Ethylenediamine

puriss. p.a., absolute, ≥99.5% (GC)

Synonym: 1,2-Diaminoethane

  • CAS Number 107-15-3

  • Linear Formula NH2CH2CH2NH2

  • Molecular Weight 60.10

  •  Beilstein Registry Number 605263

  •  EC Number 203-468-6

  •  MDL number MFCD00008204

  •  PubChem Substance ID 24845499

  •  eCl@ss 39030201

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, E-H, Puriss p.a., Puriss p.a. More...
vapor density   2.07 (vs air)
vapor pressure   10 mmHg ( 20 °C)
grade   absolute
  puriss. p.a.
assay   ≥99.5% (GC)
autoignition temp.   716 °F
expl. lim.   16 %
impurities   ≤0.5% water
evapn. residue   ≤0.05%
refractive index   n20/D 1.456
  n20/D 1.4565(lit.)
bp   118 °C(lit.)
mp   8.5 °C(lit.)
density   0.899 g/mL at 25 °C(lit.)
cation traces   Ca: ≤5 mg/kg
  Cd: ≤1 mg/kg
  Co: ≤1 mg/kg
  Cr: ≤1 mg/kg
  Cu: ≤1 mg/kg
  Fe: ≤1 mg/kg
  K: ≤20 mg/kg
  Mg: ≤1 mg/kg
  Mn: ≤1 mg/kg
  Na: ≤20 mg/kg
  Ni: ≤1 mg/kg
  Pb: ≤1 mg/kg
  Zn: ≤1 mg/kg
Gene Information   human ... FNTA(2339)

Price and Availability

Customers Also Viewed

Ethylenediamine

BioXtra

Ethylenediamine

analytical standard

Ethylenediamine

purified by redistillation, ≥99.5%

Ethylenediamine

ReagentPlus®, ≥99%

Safety & Documentation

Safety Information

Signal word 
Danger
Precautionary statements 
Hazard Codes (Europe) 
C
Risk Statements (Europe) 
Safety Statements (Europe) 
23-26-36/37/39-45
RIDADR 
UN 1604 3(8) / PGII
WGK Germany 
2
RTECS 
KH8575000
Flash Point(F) 
100.4 °F
Flash Point(C) 
38 °C

Protocols & Articles

Articles

Determination of Water Content in 1,2-Diaminoethane (Ethylene diamine) Using Karl Fischer Titration

The determination of water in 1,2-diaminoethane presents two different sorts of difficulties: 1. The compound reacts strongly alkaline and it shifts the pH of the medium in the titration vessel to an...
Keywords: Methylations, Pharmaceutical, Solvents, Titrations

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Ethylenediamine, profile of a sensitizing excipient. Zuidema J Pharm. Weekbl. Sci. 7(4), 134-40, (1985)

Loading...


Am(m)ines make the difference: organoruthenium am(m)ine complexes and their chemistry in anticancer drug development. Babak MV, Meier SM, Legin AA, et al. Chemistry 19(13), 4308-18, (2013)

Loading...


[Allergic dermatitis caused by contact with ethylenediamine]. Balato N, Cusano F, Lembo G, et al. G. Ital. Dermatol. Venereol. 122(11), 583-8, (1987)

Loading...


[A case of aminophylline hypersensitivity reaction due to ethylenediamine]. Yoshizawa A, Araki Y, Kobayashi N, et al. Arerugi. 48(11), 1206-11, (1999)

Loading...


An orally delivered small-molecule formulation with antiangiogenic and anticancer activity. Benny O Nat. Biotechnol. 26(7), 799-807, (2008)

Loading...


A sensor tip based on carbon nanotube-ink printed electrode for the dengue virus NS1 protein. Dias AC, Gomes-Filho SL, Silva MM, et al. Biosens. Bioelectron. 44, 216-21, (2013)

Loading...


Amine functionalized graphene oxide/CNT nanocomposite for ultrasensitive electrochemical detection of trinitrotoluene. Sablok K, Bhalla V, Sharma P, et al. J. Hazard. Mater. 248-249, 322-8, (2013)

Loading...


A critical reinvestigation of the TAED-activated peroxide system for low-temperature bleaching of cotton. Xu C, Long X, Du J, et al. Carbohydr. Polym. 92(1), 249-53, (2013)

Loading...


Nonfouling hydrophilic poly(ethylene glycol) engraftment strategy for PDMS/SU-8 heterogeneous microfluidic devices. Yeh PY, Zhang Z, Lin M, et al. Langmuir 28(46), 16227-36, (2012)

Loading...


A re-investigation of the Mark-Houwink-Sakurada parameters for cellulose in Cuen: a study based on size-exclusion chromatography combined with multi-angle light scattering and viscometry. Kes M and Christensen BE J. Chromatogr. A 1281, 32-7, (2013)

Loading...


Remediation of Cu(II), Ni(II), and Cr(III) ions from simulated wastewater by dendrimer/titania composites. Barakat MA, Ramadan MH, Alghamdi MA, et al. J. Environ. Manage. 117, 50-7, (2013)

Loading...


Synthesis and X-ray structures of novel macrocycles and macrobicycles containing N,N-di(pyrrolylmethyl)-N-methylamine moiety: preliminary anion binding study. Kumar R, Guchhait T, and Mani G Inorg. Chem. 51(16), 9029-38, (2012)

Loading...


Synthesis of ethylenediamine modified chitosan and evaluation for removal of divalent metal ions. Chethan PD and Vishalakshi B Carbohydr. Polym. 97(2), 530-6, (2013)

Loading...


New hyaluronic acid based brush copolymers synthesized by atom transfer radical polymerization. Pitarresi G, Fiorica C, Licciardi M, et al. Carbohydr. Polym. 92(2), 1054-63, (2013)

Loading...


Competitive binding sites of a ruthenium arene anticancer complex on oligonucleotides studied by mass spectrometry: ladder-sequencing versus top-down. Wu K, Hu W, Luo Q, et al. J. Am. Soc. Mass Spectrom. 24(3), 410-20, (2013)

Loading...


The role of the helper lipid dioleoylphosphatidylethanolamine (DOPE) for DNA transfection cooperating with a cationic lipid bearing ethylenediamine. Mochizuki S, Kanegae N, Nishina K, et al. Biochim. Biophys. Acta 1828(2), 412-8, (2013)

Loading...


Synthesis of novel octa-cationic and non-ionic 1,2-ethanediamine substituted zinc (Ⅱ) phthalocyanines and their in vitro anti-cancer activity comparison. Wang A, Zhou L, Fang K, et al. Eur. J. Med. Chem. 58, 12-21, (2012)

Loading...


Ethylenediamine diacetate (EDDA) mediated synthesis of aurones under ultrasound: their evaluation as inhibitors of SIRT1. Manjulatha K, Srinivas S, Mulakayala N, et al. Bioorg. Med. Chem. Lett. 22(19), 6160-5, (2012)

Loading...


Autofluorescence generation and elimination: a lesson from glutaraldehyde. Lee K, Choi S, Yang C, et al. Chem. Commun. (Camb.) 49(29), 3028-30, (2013)

Loading...


Synthesis and DNA interaction of ethylenediamine platinum(II) complexes linked to DNA intercalants. Duskova K, Sierra S, Fernández MJ, et al. Bioorg. Med. Chem. 20(24), 7112-8, (2012)

Loading...


Degradable poly(amidoamine) hydrogels as scaffolds for in vitro culturing of peripheral nervous system cells. Mauro N, Manfredi A, Ranucci E, et al. Macromol. Biosci. 13(3), 332-47, (2013)

Loading...


Photoluminescence of silicon quantum dots in nanospheres. Zhang Y, Han X, Zhang J, et al. Nanoscale 4(24), 7760-5, (2012)

Loading...


Synthesis and spectral studies on Pb(II) dithiocarbamate complexes containing benzyl and furfuryl groups and their use as precursors for PbS nanoparticles. Sathiyaraj E and Thirumaran S Spectrochim. Acta. A. Mol. Biomol. Spectrosc. 97, 575-81, (2012)

Loading...


Structural and functional characteristics of xenavidin, the first frog avidin from Xenopus tropicalis. Määttä JA, Helppolainen SH, Hytönen VP, et al. BMC Struct. Biol. 9, 63, (2009)

Loading...


Accessibility and selective stabilization of the principal spin states of iron by pyridyl versus phenolic ketimines: modulation of the 6A1 ↔ 2T2 ground-state transformation of the [FeN4O2]+ chromophore. Shongwe MS, Al-Zaabi UA, Al-Mjeni F, et al. Inorg. Chem. 51(15), 8241-53, (2012)

Loading...


Nitric oxide reactivity of Cu(II) complexes of tetra- and pentadentate ligands: structural influence in deciding the reduction pathway. Kumar P, Kalita A, and Mondal B Dalton Trans. 42(16), 5731-9, (2013)

Loading...


Influence of different length spacers containing enzyme conjugate on functional parameters of progesterone ELISA. Shrivastav TG, Chaube SK, Kariya KP, et al. J. Immunoassay Immunochem. 34(1), 94-108, (2013)

Loading...


Capillary electrophoresis of heparin and other glycosaminoglycans using a polyamine running electrolyte. Loegel TN, Trombley JD, Taylor RT, et al. Anal. Chim. Acta 753, 90-6, (2012)

Loading...


Platinum(II) complexes containing long-chain hydrophobic N-alkyl-diamine ligands: synthesis, characterization, molecular modeling, and cytotoxic activity. Tavares TT, Teixeira GF, Lopes CM, et al. J. Inorg. Biochem. 115, 13-9, (2012)

Loading...


Interaction of drug based binuclear mixed-ligand complexes with DNA. Mohan N Patel et al Bioorg. Med. Chem. 17, 5648-55, (2009)

Loading...


Novel pyranopyrazoles: synthesis and theoretical studies. Al-Amiery AA, Al-Bayati RI, Saed FM, et al. Molecules 17(9), 10377-89, (2012)

Loading...


Spectroscopic and electrochemical characterization of some Schiff base metal complexes containing benzoin moiety. El-Shahawi MS, Al-Jahdali MS, Bashammakh AS, et al. Spectrochim. Acta. A. Mol. Biomol. Spectrosc. 113, 459-65, (2013)

Loading...


Antitubercular activity of alpha,omega-diaminoalkanes, H2N(CH2)nNH2. Fátima M F Vergara et al Bioorg. Med. Chem. Lett. 19, 4937-8, (2009)

Loading...


Aging effects of plasma polymerized ethylenediamine (PPEDA) thin films on cell-adhesive implant coatings. Testrich H, Rebl H, Finke B, et al. Mater. Sci. Eng. C. Mater. Biol. Appl. 33(7), 3875-80, (2013)

Loading...


In vivo examination of the local inflammatory response after implantation of Ti6Al4V samples with a combined low-temperature plasma treatment using pulsed magnetron sputtering of copper and plasma-polymerized ethylenediamine. Hoene A, Patrzyk M, Walschus U, et al. J. Mater. Sci. Mater. Med. 24(3), 761-71, (2013)

Loading...


Allergic contact dermatitis to ethylenediamine. A continuing problem. Hogan DJ Dermatol. Clin. 8(1), 133-6, (1990)

Loading...


Effects of ethylenediamine in rodent models of seizure, motor coordination and anxiety. Addae JI, Walkins K, Cruickshank R, et al. Brain Res. 1473, 155-60, (2012)

Loading...


Cytotoxic activity of N, N'-Bis (2-hydroxybenzyl) ethylenediamine derivatives in human cancer cell lines. Musa MA, Badisa VL, and Latinwo LM Anticancer Res. 34(4), 1601-7, (2014)

Loading...


Merck 14,3795

Beil. 4,IV,1166

Fieser 1,372 / Fieser 4,231 / Fieser 10,297 / Fieser 12,338 / Fieser 13,157

Aldrich MSDS 1, 902:D / Corp MSDS 1 (1), 1604:B / FT-IR 2 (1), 453:C / FT-IR 1 (1), 289:C / FT-NMR 1 (1), 463:B / IR-Spectra (2), 154:A / IR-Spectra (3), 171:A / NMR-Reference 2 (1), 248:A / RegBook 1 (1), 309:A / Sax 6, 1303 / Sigma FT-IR 1 (2), 547:A / Structure Index 1, 42:A:3 / Vapor Phase 3, 374:D

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?