• USA Home
  • 03701 - Ethylenediaminetetraacetic acid tetrasodium salt hydrate

EMAIL THIS PAGE TO A FRIEND

03701 Fluka

Ethylenediaminetetraacetic acid tetrasodium salt hydrate

≥97.0% (KT)

Synonym: EDTA tetrasodium salt, Edathamil, Tetrasodium ethylenediaminetetraacetate hydrate

  • CAS Number 194491-31-1

  • Linear Formula (NaOOCCH2)2NCH2CH2N(CH2COONa)2 · xH2O

  • Molecular Weight 380.17 (anhydrous basis)

  •  Beilstein Registry Number 3861753

  •  EC Number 200-573-9

  •  MDL number MFCD00150025

  •  PubChem Substance ID 24845563

Purchase

Properties

Related Categories Chelation/Complexation Compounds, Chemical Synthesis, EDTA Derivatives, Synthetic Reagents
assay   ≥97.0% (KT)

Description

Preparation Note

Soluble in water at room temperature up to 1.45 M. The pH of this solution will be approximately 10 to 11 and will be rather viscous.

Application

EDTA is an inhibitor of metalloproteases, at effective concentrations of 1-10 mM. EDTA acts as a chelator of the zinc ion in the active site of metalloproteases, and can also inhibit other metal ion-dependent proteases such as calcium-dependent cysteine proteases. For use as an anticoagulant, disodium or tripotassium salts of EDTA are most commonly used. The optimal concentration is 1.5 mg per ml of blood.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
Hazard Codes (Europe) 
Xn
Risk Statements (Europe) 
Safety Statements (Europe) 
26-39-46
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Merck 14,3517

Beil. 4,IV,2451

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?