• USA Home
  • 03854 - Creosote from beechwood tar

EMAIL THIS PAGE TO A FRIEND

03854 Fluka

Creosote from beechwood tar

Synonym: Beechwood creosote

Purchase

Properties

Related Categories Cell Biology, Nutrition Research, Phytochemicals by Chemical Classification, Plant Extracts
ign. residue   ≤0.01% (as SO4)
bp   200-220 °C(lit.)
transition temp   solidification point <−20 °C
density   1.09 g/mL at 20 °C

Price and Availability

Customers Also Viewed

2-<WBR>Methoxy-<WBR>4-<WBR>methylphenol

≥98%, FG

Nordihydroguaiaretic acid

≥90% (HPLC), from Larrea divaricata (creosote bush)

<I>o</I>-Cresol
C85700o-Cresol

ReagentPlus®, ≥99%

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard Codes (Europe) 
T
Risk Statements (Europe) 
Safety Statements (Europe) 
26-36/37/39-45-61
RIDADR 
UN 2922 6.1(8) / PGII
WGK Germany 
3
Flash Point(F) 
165.2 °F
Flash Point(C) 
74 °C

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Sorption of native polyaromatic hydrocarbons (PAH) to black carbon and amended activated carbon in soil. Brändli RC, Hartnik T, Henriksen T, et al. Chemosphere 73(11), 1805-10, (2008)

Loading...


Spatial patterns of microbial diversity and activity in an aged creosote-contaminated site. Mukherjee S, Juottonen H, Siivonen P, et al. ISME J. 8(10), 2131-42, (2014)

Loading...


Expression of biotransformation genes in woodrat (Neotoma) herbivores on novel and ancestral diets: identification of candidate genes responsible for dietary shifts. Magnanou E, Malenke JR, and Dearing MD Mol. Ecol. 18(11), 2401-14, (2009)

Loading...


[A historical review of the therapeutic use of wood creosote. Part II: Original plant source of crude drug wood creosote]. Moriguchi N, Sato A, Shibata T, et al. Yakushigaku Zasshi. 46(1), 13-20, (2011)

Loading...


Endodontic microorganism susceptibility by direct contact test. Pérez SB, Tejerina DP, Pérez Tito RI, et al. Acta Odontol. Latinoam. 21(2), 169-73, (2008)

Loading...


Microbial diversity and PAH catabolic genes tracking spatial heterogeneity of PAH concentrations. Bengtsson G, Törneman N, De Lipthay JR, et al. Microb. Ecol. 65(1), 91-100, (2013)

Loading...


A spatial approach to environmental risk assessment of PAH contamination. Bengtsson G and Törneman N Risk Anal. 29(1), 48-61, (2009)

Loading...


Removal of organic matter from a variety of water matrices by UV photolysis and UV/H2O2 method. Vilhunen S, Vilve M, Vepsäläinen M, et al. J. Hazard. Mater. 179(1-3), 776-82, (2010)

Loading...


Electrochemical degradation of polycyclic aromatic hydrocarbons in creosote solution using ruthenium oxide on titanium expanded mesh anode. Tran LH, Drogui P, Mercier G, et al. J. Hazard. Mater. 164(2-3), 1118-29, (2009)

Loading...


Quantitative source apportionment of PAHs in sediments of Little Menomonee River, Wisconsin: weathered creosote versus urban background. Stout SA and Graan TP Environ. Sci. Technol. 44(8), 2932-9, (2010)

Loading...


Oxidation by Fenton's reagent combined with biological treatment applied to a creosote-comtaminated soil. Valderrama C, Alessandri R, Aunola T, et al. J. Hazard. Mater. 166(2-3), 594-602, (2009)

Loading...


Analysis of phenanthrene and benzo[a]pyrene tetraol enantiomers in human urine: relevance to the bay region diol epoxide hypothesis of benzo[a]pyrene carcinogenesis and to biomarker studies. Hecht SS, Carmella SG, Villalta PW, et al. Chem. Res. Toxicol. 23(5), 900-8, (2010)

Loading...


Bioavailability of residual polycyclic aromatic hydrocarbons following enhanced natural attenuation of creosote-contaminated soil. Juhasz AL, Smith E, Waller N, et al. Environ. Pollut. 158(2), 585-91, (2010)

Loading...


Spatial uncoupling of biodegradation, soil respiration, and PAH concentration in a creosote contaminated soil. Bengtsson G, Törneman N, and Yang X Environ. Pollut. 158(9), 2865-71, (2010)

Loading...


Effects of benzo[a]pyrene on mitochondrial and nuclear DNA damage in Atlantic killifish (Fundulus heteroclitus) from a creosote-contaminated and reference site. Jung D, Cho Y, Collins LB, et al. Aquat. Toxicol. 95(1), 44-51, (2009)

Loading...


Microbial populations related to PAH biodegradation in an aged biostimulated creosote-contaminated soil. Lladó S, Jiménez N, Viñas M, et al. Biodegradation 20(5), 593-601, (2009)

Loading...


Degradation of PAH in a creosote-contaminated soil. A comparison between the effects of willows (Salix viminalis), wheat straw and a nonionic surfactant. Hultgren J, Pizzul L, Castillo Mdel P, et al. Int. J. Phytoremediation 12(1), 54-66, (2010)

Loading...


Self-sustaining smoldering combustion for NAPL remediation: laboratory evaluation of process sensitivity to key parameters. Pironi P, Switzer C, Gerhard JI, et al. Environ. Sci. Technol. 45(7), 2980-6, (2011)

Loading...


An integrated assessment of sediment remediation in a midwestern U.S. stream using sediment chemistry, water quality, bioassessment, and fish biomarkers. Meier JR, Snyder S, Sigler V, et al. Environ. Toxicol. Chem. 32(3), 653-61, (2013)

Loading...


Functional genes reveal the intrinsic PAH biodegradation potential in creosote-contaminated groundwater following in situ biostimulation. Nyyssönen M, Kapanen A, Piskonen R, et al. Appl. Microbiol. Biotechnol. 84(1), 169-82, (2009)

Loading...


A versatile cryo-focussing flow switching gas chromatography inlet for trace analysis of intractable compounds. Apps P and Mmualefe L J. Chromatogr. A 1257, 171-88, (2012)

Loading...


Biotransformation enzyme expression in the nasal epithelium of woodrats. Skopec MM, Hale A, Torregrossa AM, et al. Comp. Biochem. Physiol. C. Toxicol. Pharmacol. 157(1), 72-9, (2013)

Loading...


Two cases of gastric Anisakiasis for which oral administration of a medicine containing wood creosote (Seirogan) was effective. Sekimoto M, Nagano H, Fujiwara Y, et al. Hepatogastroenterology. 58(109), 1252-4, (2011)

Loading...


Diversity of fungi in creosote-treated crosstie wastes and their resistance to polycyclic aromatic hydrocarbons. Kim MJ, Lee H, Choi YS, et al. Antonie van Leeuwenhoek 97(4), 377-87, (2010)

Loading...


A comparative toxicity assessment of materials used in aquatic construction. Lalonde BA, Ernst W, Julien G, et al. Arch. Environ. Contam. Toxicol. 61(3), 368-75, (2011)

Loading...


A microarray's view of life in the desert: adding a powerful evolutionary genomics tool to the packrat's midden. Matocq MD Mol. Ecol. 18(11), 2310-2, (2009)

Loading...


Merck 14,2573

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?