Synonym: Standard - Nutrient Broth
MDL number MFCD01867738
Popular Documents: Datasheet (PDF)
| Related Categories | All Dairy Media (alphabetical), All Liquid Media/Broths, Bacillus, Bacteria (General Media), Candida, |
| grade | for microbiology |
| shelf life | (limited shelf life, expiry date on the label) |
| composition | meat extract, 3 g/L |
| meat peptone, 5 g/L | |
| final pH | 7.0±0.2 (25 °C) |
| suitability | nonselective for Bacillus |
| nonselective for Bacteria (General Media) |
Universal broth for the cultivation of less critical microorganisms.
Customers Also Viewed
Microbiologically tested
No. 1, for microbiology
No. 2, for microbiology
No. 3, for microbiology
for microbiology
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?