03884 Fluka

Cetyl alcohol

Ph Eur

DOWNLOAD MSDS (PDF)

Synonym: 1-Hexadecanol, Alcohol cetylicus, Palmityl alcohol

  • CAS Number 36653-82-4

  • Linear Formula CH3(CH2)15OH

  • Molecular Weight 242.44

  •  Beilstein Registry Number 1748475

  •  EC Number 253-149-0

  •  MDL number MFCD00004760

  •  PubChem Substance ID 24845613

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Pharmacopoeia, Pharmacopoeia A-Z More...
grade   Ph Eur
pharmacopeia   testing & handling conforms to Pharmacopeia
bp   190 °C/15 mmHg(lit.)

Price and Availability

Customers Also Viewed

1-Hexadecanol

ReagentPlus®, 99%

Cetyl alcohol

Selectophore, ≥99.0%

Cetyl alcohol

analytical standard

Cetyl Alcohol

pharmaceutical secondary standard; traceable to USP and PhEur

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?