EMAIL THIS PAGE TO A FRIEND

03892 Fluka

Hayem’s solution

for counting (of erythrocytes)

Purchase

Properties

grade   for counting (of erythrocytes)
impurities   mercuric chloride

Price and Availability

Customers Also Viewed

for the determination of hemoglobin

for microscopy (for counting of Leukozytes)

Safety & Documentation

Safety Information

Symbol 
GHS09  GHS09
Hazard statements 
Precautionary statements 
Hazard Codes (Europe) 
Risk Statements (Europe) 
Safety Statements (Europe) 
36/37-61
RIDADR 
UN 2024 6.1 / PGII
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?