EMAIL THIS PAGE TO A FRIEND

04390 Fluka

2-Butanone peroxide

technical

Synonym: Ethyl methyl ketone peroxide, Methyl ethyl ketone peroxide

  • CAS Number 1338-23-4

  • Linear Formula C2H5C(OOH)(CH3)OOC(OOH)(CH3)C2H5

  • Molecular Weight 210.22

  •  Beilstein Registry Number 1759757

  •  MDL number MFCD00042641

  •  PubChem Substance ID 24845732

  • Popular Documents:  FTIR (PDF)  

Purchase

Properties

grade   technical
contains   50-60% diacetone alcohol + phthalic acid ester as plasticizer
refractive index   n20/D 1.455
density   1.053 g/mL at 20 °C(lit.)
storage temp.   2-8°C

Description

Application

Reactant for:
• Studies of the adiabatic runaway reaction of Me Et Ketone peroxide1
• Quantitation of metal ions in archaeological glass via abrasive stripping square-wave voltammetry2
• Imaging of hydrogen peroxide and hydrogen peroxide-scavenging substances by photon emission3
• Synthesis of oligonucleotides via phosphoramidite method4
• Oxidation of nucleoside phosphites into phosphates5
• Comparing the relative effectiveness of human plasma glutathione peroxidase as a catalyst for reduction of hydroperoxidases6

Analyte for direct electrochemical catalystis of immobilized Hb in an ethanol-water mixture7

Price and Availability

Customers Also Viewed

2-Butanone peroxide

technical

Luperox<SUP>®</SUP> DDM-9, 2-Butanone peroxide solution

~35 wt. % in 2,2,4-trimethyl-1,3-pentanediol diisobutyrate

Luperox<SUP>®</SUP> DHD-9, 2-Butanone peroxide solution

~32 wt. % in phthalate-free plasticizer mixture

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Precautionary statements 
Hazard Codes (Europe) 
Risk Statements (Europe) 
Safety Statements (Europe) 
53-3/7-14-26-36/37/39-45-61
RIDADR 
UN 3105A5 5.2 / PGII
WGK Germany 
3
Flash Point(F) 
138.2 °F
Flash Point(C) 
59 °C

Protocols & Articles

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?