CAS Number 259853-21-9
Empirical Formula (Hill Notation) C14H10ClNO3
Molecular Weight 275.69
MDL number MFCD06798159
| Related Categories | Aryl, Building Blocks, C13 to C37+, Chemical Synthesis, Halogenated Hydrocarbons, |
| grade | purum |
| assay | ≥97.0% (T) |
| storage temp. | 2-8°C |
Visit our Sensor Applications portal to learn more.
Determination of Fe(III) in rock, direct measurement or as indicator electrode in complexometric titrations1
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?