04621 Fluka

Oxalate standard for IC

analytical standard, 1.000 g/L in H2O

DOWNLOAD MSDS (PDF)

Synonym: Oxalic acid standard solution

  • CAS Number 144-62-7

  • Linear Formula HO2CCO2H

  • Molecular Weight 90.03

  •  Beilstein Registry Number 3679436

  •  EC Number 231-791-2

  •  MDL number MFCD00002573

  •  PubChem Substance ID 24845757

Properties

Related Categories Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Anionic Solutions, Bioactive Small Molecules,
grade   analytical standard
shelf life   (limited shelf life, expiry date on the label)
concentration   1.000 g/L in H2O
Gene Information   human ... SRC(6714)

Description

Other Notes

Aqueous solutions of oxalate in 0.1% benzoic acid.

Preparation Note

prepared with oxalic acid and H2O

General description

filtered through a 0.45 μm membrane filter

Price and Availability

Customers Also Viewed

1000 mg/L, analytical standard

Formate standard for IC

1.000 g/L in H2O, analytical standard

analytical standard

TraceCERT®, 1000 mg/L oxalate in H2O

Oxalic acid

puriss. p.a., anhydrous, ≥99.0% (RT)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?