04991 Fluka

2,6-Lutidine

analytical standard

DOWNLOAD MSDS (PDF)

Synonym: 2,6-Dimethylpyridine

  • CAS Number 108-48-5

  • Empirical Formula (Hill Notation) C7H9N

  • Molecular Weight 107.15

  •  Beilstein Registry Number 105690

  •  EC Number 203-587-3

  •  MDL number MFCD00006345

  •  PubChem Substance ID 24845651

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories All Alphabetically Sorted, Analytical Standards, Analytical/Chromatography, Chemical Class, Chromatography,
grade   analytical standard
assay   ≥99.7% (GC)
shelf life   (limited shelf life, expiry date on the label)
refractive index   n20/D 1.497(lit.)
  n20/D 1.499
bp   143-145 °C(lit.)
mp   −6 °C(lit.)
density   0.92 g/mL at 25 °C(lit.)

Price and Availability

Customers Also Viewed

2,6-Lutidine

purified by redistillation, ≥99%

2,6-Lutidine

ReagentPlus®, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?