05428 Fluka

Viscosity and Density Standard

S20, UKAS ISO/IEC17025 and ISO Guide 34 certified, viscosity 28.42 mPa.s (25 °C)

DOWNLOAD MSDS (PDF)
  •  EC Number 232-455-8

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Application Areas and Market Segments, Petrochemistry, Pharma,
shelf life   (limited shelf life, expiry date on the label)
mfr. no.   Paragon Scientific Ltd.
viscosity   28.42 mPa.s(25 °C)
density   0.8474 g/mL at 25 °C

Price and Availability

Customers Also Viewed

S3, UKAS ISO/IEC17025 and ISO Guide 34 certified, viscosity 3.251 mPa.s (25 °C)

S6, UKAS ISO/IEC17025 and ISO Guide 34 certified, viscosity 7.245 mPa.s (25 °C)

D10, UKAS ISO/IEC17025 and ISO Guide 34 certified, viscosity 10.07 mPa.s (25 °C)

N10, UKAS ISO/IEC17025 and ISO Guide 34 certified, viscosity 14.43 mPa.s (25 °C)

N35, UKAS ISO/IEC17025 and ISO Guide 34 certified, viscosity 55.43 mPa.s (25 °C)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?