Synonym:
Fame 22:6n-3,
Methyl DHA,
Methyl all-cis-
CAS Number 2566-90-7
Empirical Formula (Hill Notation) C23H34O2
Molecular Weight 342.51
MDL number MFCD00673450
| Related Categories | Analytical Standards, Analytical/Chromatography, Building Blocks, C12 to C63, Carbonyl Compounds, |
| grade | analytical standard |
| assay | ≥98.5% (GC) |
| shelf life | (limited shelf life, expiry date on the label) |
Bottomless glass bottle. Contents are inside inserted fused cone.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?