CAS Number 52918-63-5
Empirical Formula (Hill Notation) C22H19Br2NO3
Molecular Weight 505.20
Beilstein Registry Number 6746312
EC Number 258-256-6
MDL number MFCD00870122
| Related Categories | Analytical Standards, Analytical/Chromatography, CRM for chromatography, Certified Reference Materials (CRMs), Organic CRMs Alphabetical, |
| grade | certified reference material |
| TraceCERT® | |
| shelf life | (limited shelf life, expiry date on the label) |
| storage temp. | 2-8°C |
Certified content by quantitative NMR incl. uncertainty and expiry are given in the certificate.
Download your certificate at: http://www.sigma-aldrich.
Bottomless glass bottle. Contents are inside inserted fused cone.
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?