06220 Fluka

Aluminum chloride

puriss. p.a., anhydrous, crystallized, ≥99.0% (AT)

DOWNLOAD MSDS (PDF)
  • CAS Number 7446-70-0

  • Linear Formula AlCl3

  • Molecular Weight 133.34

  •  EC Number 231-208-1

  •  MDL number MFCD00003422

  •  PubChem Substance ID 24845807

Properties

Related Categories A-B, Puriss p.a., Aluminum, Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography,
vapor pressure   1 mmHg ( 100 °C)
grade   puriss. p.a.
assay   ≥99.0% (AT)
quality   crystallized
mp   190 °C(lit.)
anion traces   sulfate (SO42-): ≤100 mg/kg
cation traces   Ca: ≤20 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤10 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤10 mg/kg
  Mg: ≤10 mg/kg
  Na: ≤10 mg/kg
  Ni: ≤10 mg/kg
  Pb: ≤5 mg/kg
  Zn: ≤10 mg/kg

Description

Caution

may contain some grey particles

Features and Benefits

The vapor-phase co-reductions with other metal halides by hydrogen results in finely divided intermetallics with applications as structural materials or compounds with useful thermoelectric, magnetic, and oxidation-resistance properties.1 Aluminum nitride can be synthesized from the elemnts with AlCl3 added to help facilitate crystallization.2

Price and Availability

Customers Also Viewed

Aluminum chloride

anhydrous, powder, 99.999% trace metals basis

Aluminum chloride

anhydrous, powder, 99.99% trace metals basis

Aluminum chloride

meets EP, BP, USP testing specifications

Aluminum chloride

99.99% trace metals basis

Aluminum chloride

ReagentPlus®, anhydrous, ≥99.9% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?