06231 Fluka

Aluminum chloride hexahydrate

Ph Eur

DOWNLOAD MSDS (PDF)

Synonym: Aluminii chloridum hexahydricum

  • CAS Number 7784-13-6

  • Empirical Formula (Hill Notation) AlCl3 · 6H2O

  • Molecular Weight 241.43

  •  EC Number 231-208-1

  •  MDL number MFCD00149134

Properties

Related Categories Aluminum, Catalysis and Inorganic Chemistry, Chemical Synthesis, Lewis Acids, Synthetic Reagents More...
grade   Ph Eur
vapor pressure   1 mmHg ( 100 °C)
pharmacopeia   testing & handling conforms to Pharmacopeia

Price and Availability

Customers Also Viewed

Aluminum chloride

meets EP, BP, USP testing specifications

Aluminum chloride hexahydrate

purum p.a., crystallized, ≥99.0% (AT)

Aluminum chloride hexahydrate

puriss., meets analytical specification of Ph. Eur., BP, USP, 97.0-101.0%

Aluminum chloride hexahydrate

ReagentPlus®, 99%

Aluminum chloride hydrate

99.999% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?