Synonym:
1,3,6,7-
CAS Number 4773-96-0
Empirical Formula (Hill Notation) C19H18O11
Molecular Weight 422.34
MDL number MFCD00075656
| Related Categories | Alphabetical, Analytical Standards, Analytical/Chromatography, Chromatography, Flavonoids, |
| grade | analytical standard |
| assay | ≥98.0% (TLC) |
| shelf life | (limited shelf life, expiry date on the label) |
| storage temp. | 2-8°C |
Bottomless glass bottle. Contents are inside inserted fused cone.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?