• USA Home
  • 06391 - Ammonium chloride 20 mM – Samarium(III) chloride 1 mM – TRIS Buffer pH 8.0; 50 mM – Ethanol 20% solution

EMAIL THIS PAGE TO A FRIEND

06391 Fluka

Ammonium chloride 20 mM – Samarium(III) chloride 1 mM – TRIS Buffer pH 8.0; 50 mM – Ethanol 20% solution

Synonym: Crystallization solution 33/Fluka kit no 18839

Purchase

Properties

pH   8.0±0.2 ( in neat)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
Risk Statements (Europe) 
10
RIDADR 
UN 1170C 3 / PGIII
WGK Germany 
1
Flash Point(F) 
95 °F
Flash Point(C) 
35 °C

Documents

Certificate of Analysis

Protocols & Articles

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?