Synonym: Amberlite® IR120 hydrogen form
CAS Number 39389-20-3
MDL number MFCD00132707
| Related Categories | Amberlite, Amberlite Strong Cation Exchangers, Analytical/Chromatography, Cation, Cation Exchange Media, |
| vapor density | >1 (vs air) |
| vapor pressure | 17 mmHg ( 20 °C) |
| autoignition temp. | 800 °F |
| limit | 121 °C max. temp. |
| cross-linkage | 8% |
| moisture | 53-58% |
| loss | ~50% loss on drying, 110 °C |
Strongly acidic cation exchange resin suitable for a wide variety of chemical process applications, removal of amino acids at low pH, USP potassium methods, etc..
Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow
Customers Also Viewed
H+-form, strongly acidic, 200-400 mesh
hydrogen form
Na+-form
strongly acidic, cation exchanger, dry
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?