• USA Home
  • 06728 - Hydrogen Sulfide Test Strips

EMAIL THIS PAGE TO A FRIEND

06728 Fluka

Hydrogen Sulfide Test Strips

for microbiology

Synonym: H2S test strips, Lead acetate test strips

Purchase

Properties

Related Categories Discs, Strips and Cards, Identification Tests & Reagents, Microbiology More...
grade   for microbiology
shelf life   limited shelf life, expiry date on the label
storage temp.   2-8°C

Description

General description

These strips are used for detection of hydrogen sulfide production by microorganisms. Hydrogen sulfide can be produced in small amounts from sulfur containing amino acids like cysteine by a large number of bacteria in carbohydrate media. Hydrogen sulfide produces on contact with lead acetate a black preciptate, indicated by a visible black-colored reaction on the hydrogen sulfide paper strip. The lead acetate procedure is more sensitive than any other method for detecting hydrogen sulfide production.

Packaging

Sterile filter paper strips (25 pcs, diameter 7 mm) impregnated with lead acetate

Price and Availability

Customers Also Viewed

reag. DAB7

radiello<SUP>™</SUP> Cartridge Adsorbents

for sampling Hydrogen Sulfide (H2S), matrix microporous PE coated with zinc acetate, pk of 20

Whatman<SUP>®</SUP> indicator papers

Specialized Test Papers, Lead Acetate

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Hazard Codes (Europe) 
T,N
Risk Statements (Europe) 
Safety Statements (Europe) 
53-36/37-45-61
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Identification of Microorganisms Based on Color

For microbiologists the most fundamental stain was developed in 1884 by the Danish bacteriologist Hans Christian Gram. The Gram Stain allows the determination of morphology, dividing bacteria into tw...
Jvo Siegrist
Microbiology Focus Edition 1.4, Product Manager Microbiology.... ivo.siegrist@sial.com
Keywords: Antibiotics, Enzyme activity, Fermentation, Gram stain, Help, Indicators, Reductions

On the trail of Campylobacter

The Campylobacter is one of the leading causes of human gastroenteritis. Common Campylobacter species C. jejuni, C. coli, and C. lari are responsible for most cases of campylobacteriosis1,2. However,...
Jvo Siegrist
AnalytiX Volume 8 Issue 1
Keywords: Antibiotics, Cancer, Citric Acid Cycle, Gram stain, Metabolism, Microbiology, Scanning electron microscopy

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?