06931 Fluka

Pyrogallol

analytical standard

DOWNLOAD MSDS (PDF)

Synonym: 1,2,3-Trihydroxybenzene

  • CAS Number 87-66-1

  • Linear Formula C6H3(OH)3

  • Molecular Weight 126.11

  •  Beilstein Registry Number 907431

  •  EC Number 201-762-9

  •  MDL number MFCD00002192

Properties

Related Categories Alcohols and Phenols, Alphabetical, Analytical Standards, Analytical/Chromatography, Betula,
grade   analytical standard
vapor density   4.4 (vs air)
vapor pressure   10 mmHg ( 167.7 °C)
assay   ≥98.0% (HPLC)
shelf life   (limited shelf life, expiry date on the label)
impurities   ≤0.5% water
bp   309 °C(lit.)
mp   132-134 °C
  133-134 °C(lit.)
storage temp.   2-8°C

Description

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Customers Also Viewed

Pyrogallol

≥98% (HPLC)

Pyrogallol

puriss., ACS reagent, ≥99%

Pyrogallol

purum, ≥98.0% (HPLC)

Pyrogallol

ACS reagent

Pyrogallol solution

~25% in H2O, for the absorption of oxygen in gas analysis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?