07012 Fluka

m-FC Agar modified for Klebsiella, Vegitone

for microbiology

DOWNLOAD MSDS (PDF)

Synonym: Vegitone m-FC Agar modified for Klebsiella, m-Fecal Coliform Agar modified for Klebsiella, Vegitone

Properties

Related Categories A - M, All Food Control Media (alphabetical), All Solid Media/Agars, Enterobacter, Klebsiella,
grade   for microbiology
shelf life   (limited shelf life, expiry date on the label)
composition   agar, 15.0 g/L
  aniline blue, 0.1 g/L
  inositol, 10.0 g/L
  proteose peptone (vegetable), 5.0 g/L
  sodium chloride, 5.0 g/L
  synthetic detergent, 1.5 g/L
  tryptose (vegetable), 10.0 g/L
  yeast extract, 3.0 g/L
final pH   7.4±0.2 (25 °C)
suitability   selective and differential for Enterobacter
  selective and differential for Klebsiella

Description

Application

This M-FC Agar is used for rapid enumeration of Klebsiella using membrane filter technique. It is free of animal derived material.

Price and Availability

Customers Also Viewed

for microbiology

for microbiology

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?