07097 Fluka

Aminobenzyl-EDTA

≥90% (CHN)

DOWNLOAD MSDS (PDF)

Synonym: 1-(4-Aminobenzyl)ethylenediamine-N,N,N′,N′-tetra­acetic acid

  • CAS Number 84256-90-6

  • Empirical Formula (Hill Notation) C17H23N3O8

  • Molecular Weight 397.38

  •  MDL number MFCD00236410

  •  PubChem Substance ID 24846011

Properties

assay   ≥90% (CHN)
storage temp.   −20°C

Description

Application

Used to incorporate aminobenzyl-EDTA into peptide by reacting with activated carboxylates such as succinimidylester, p-nitrophenylester, and acid chlorides.

Aminobenzyl-EDTA has been used to develop an ELISA test for the detection of mercury in water. Aminobenzyl-EDTA has also been used in a study to provide a new class of affinity cleaving reagents that can be directed against protein and nucleic acid targets.

Other Notes

Reagent for labeling biomolecules with EDTA1

Price and Availability

Customers Also Viewed

Ethylenediaminetetraacetic acid

BioUltra, anhydrous, ≥99% (titration)

Ethylenediaminetetraacetic acid

anhydrous, crystalline, BioReagent, suitable for cell culture

Ethylenediaminetetraacetic acid

99.995% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?