• USA Home
  • 08091 - SDS Gel Preparation Kit

EMAIL THIS PAGE TO A FRIEND

08091 Fluka

SDS Gel Preparation Kit

Purchase

Properties

Related Categories Gel Casting Reagents, Molecular Biology, Protein Electrophoresis, Proteomics, SDS-PAGE More...
impurities   acrylamide
  bisacrylamide
storage temp.   2-8°C

Description

Components

Components:
250 mL Separation Gel, 3× Concentrate (Fluka 14946)
100 mL Stacking Gel, 5× Concentrate (Fluka 19505)
250 mL 30% Acrylamide/0.8% Bisacrylamide Solution (Fluka 06867)
3x 250 mL Buffer Concentrate, 10× Concentrate (Fluka 14393)
30 mL TEMED 10% (Fluka 07654)
30 mL Sample Incubation Buffer Concentrate
2× Concentrated Persulfate Solution

Application

Ready to use solutions for ~50 SDS-PAGE gels (gel size 8x8 cm, 15 ml)

Price and Availability

TruPAGE™ Pre-cast Polyacrylamide Gels


TruPAGE™ Pre-cast Polyacrylamide GelsTruPAGE Gels run true every time!

  • . 2-Year Shelf Life
  • . Superior Band Resolution
  • . Strong & Tear Resistant
  • . Higher Capacity Wells


Safety & Documentation

Safety Information

Signal word 
Danger
Precautionary statements 
Hazard Codes (Europe) 
O,T
Risk Statements (Europe) 
Safety Statements (Europe) 
53-23-26-36/37/39-45
RIDADR 
UN 3316 9
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?