Synonym:
(−)-cis-3,3′,4′,5,5′,7-Hexahydroxyflavane,
(−)-cis-
CAS Number 970-74-1
Empirical Formula (Hill Notation) C15H14O7
Molecular Weight 306.27
MDL number MFCD00075939
PubChem Substance ID 24846148
| Related Categories | Alcohols, Alphabetical, Analytical Standards, Analytical/Chromatography, Aromatic, |
| grade | analytical standard |
| assay | ≥90% (HPLC) |
| Gene Information | human ... CYP1A2(1544) |
Green tea polyphenol. Anticancer agent. Lower antioxidant activity than EGCG.
Ginkgo (Ginkgo biloba) Plant Profile: bioactives, mechanism of action, references
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?