08253 Fluka

Copper(I) chloride

0.1-0.5 mm particle size, ≥97.0%

DOWNLOAD MSDS (PDF)

Synonym: Copper monochloride, Cuprous chloride

  • CAS Number 7758-89-6

  • Linear Formula CuCl

  • Molecular Weight 99.00

  •  EC Number 231-842-9

  •  MDL number MFCD00010971

  •  PubChem Substance ID 24846153

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Copper More...
vapor pressure   1.3 mmHg ( 546 °C)
assay   ≥97.0%
particle size   0.1-0.5 mm
bp   1490 °C(lit.)
mp   430 °C(lit.)

Description

Application

Shows unique character as an initiator of radical reactions such as the hydrostannation of α,β-unsaturated ketones.1

Price and Availability

Customers Also Viewed

Copper(I) chloride

reagent grade, 97%

Copper(I) chloride

purum, ≥97.0% (RT)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?