08421 Fluka

Benzyl alcohol

analytical standard

DOWNLOAD MSDS (PDF)

Synonym: Benzenemethanol

Properties

Related Categories A - D, A-B, Alcohols, Alcohols and Phenols, Alpha Sort,
grade   analytical standard
vapor density   3.7 (vs air)
vapor pressure   13.3 mmHg ( 100 °C)
  3.75 mmHg ( 77 °C)
assay   ≥99.5% (GC)
autoignition temp.   817 °F
shelf life   (limited shelf life, expiry date on the label)
refractive index   n20/D 1.539(lit.)
  n20/D 1.541
bp   203-205 °C(lit.)
mp   −16-−13 °C(lit.)
density   1.045 g/mL at 25 °C(lit.)

Price and Availability

Proficiency Testing Samples from RTC


PT Samples from RTCRTC has been producing environmental LPTP for more than 20 years and send out over 20,000 PT samples a year, throughout
the world.





Customers Also Viewed

Benzaldehyde

analytical standard

Benzyl alcohol

anhydrous, 99.8%

Benzyl alcohol

puriss., meets analytical specification of Ph. Eur., BP, NF, 99-100.5% (GC)

Benzyl alcohol

certified reference material, TraceCERT®

Benzyl alcohol

ACS reagent, ≥99.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?