Synonym: PEG tBu propionate (n=12)
Empirical Formula (Hill Notation) C31H62O15
Molecular Weight 674.81
MDL number MFCD06201003
PubChem Substance ID 24846184
| Related Categories | Chemical Biology, Chemical Synthesis, High Oligomer Purity PEG, PEGylation |
| assay | ≥95% (oligomer purity) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?