08670 Fluka

Viscosity and Density Standard

D1000, UKAS ISO/IEC17025 and ISO Guide 34 certified, viscosity 1010 mPa.s (25 °C)

DOWNLOAD MSDS (PDF)

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Application Areas and Market Segments, Petrochemistry, Pharma,
shelf life   (limited shelf life, expiry date on the label)
mfr. no.   Paragon Scientific Ltd.
viscosity   1010 mPa.s(25 °C)
density   0.8715 g/mL at 25 °C

Price and Availability

Customers Also Viewed

D10, UKAS ISO/IEC17025 and ISO Guide 34 certified, viscosity 10.07 mPa.s (25 °C)

S60, UKAS ISO/IEC17025 and ISO Guide 34 certified, viscosity 101.7 mPa.s (25 °C)

D500, UKAS ISO/IEC17025 and ISO Guide 34 certified, viscosity 503 mPa.s (25 °C)

N350, UKAS ISO/IEC17025 and ISO Guide 34 certified, viscosity 740.3 mPa.s (25 °C)

N1000, UKAS ISO/IEC17025 and ISO Guide 34 certified, viscosity 2618 mPa.s (25 °C)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?