MDL number MFCD00003892
PubChem Substance ID 24846297
| Related Categories | Other Stains for Electrophoresis, Protein Electrophoresis, Protein Stains and Staining Reagents, Proteomics |
Live Chat and Frequently Asked Questions
are available for this Product.
Non-toxic Reagents Systems
With conventional electrophoresis reagents, there is always a risk to health. By contrast, the reagents in the ATX reagent system are non-toxic, environmentally friendly. Conventional reagents contain various toxic substances: buffer solutions contain barbituric acid, classified as a narcotic according to the Dangerous Drugs Act; staining solutions contain methanol and acetic acid which irritate the skin and airways; clarifying solutions contain methanol and dioxan isobutanol, a carcinogenic, inflammable substance which is classified as hazardous waste.
Stop using these toxic substances. For your electrophoresis, start using the non-toxic ATX reagent system and protect yourself and the environment against permanent damage.
The advantages are clear. ATX reagents are:
• non-toxic and non-irritant to skin and airways
• environmentally friendly
• disposable with regular waste water
• odorless
• non-inflammable
• stable at room temperature
• specially coordinated with ATX micro-solid strips
• available as ready-to-use solutions or as concentrates
• guaranteed to give excellent, reproducible results
6 bottles for the preparation of 250 mL ready-to-use-solution
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?