09286 Fluka

Thiophenol

analytical standard

DOWNLOAD MSDS (PDF)

Synonym: Benzenethiol, Phenyl mercaptan

  • CAS Number 108-98-5

  • Linear Formula C6H5SH

  • Molecular Weight 110.18

  •  Beilstein Registry Number 506523

  •  EC Number 203-635-3

  •  MDL number MFCD00004826

  •  PubChem Substance ID 24846302

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories All Alphabetically Sorted, Alpha Sort, Analytical Standards, Analytical/Chromatography, Carbamates,
grade   analytical standard
vapor density   3.8 (vs air)
vapor pressure   1.4 mmHg ( 20 °C)
shelf life   (limited shelf life, expiry date on the label)
refractive index   n20/D 1.588(lit.)
bp   169 °C(lit.)
mp   −15 °C(lit.)
density   1.073 g/mL at 25 °C(lit.)

Price and Availability

Proficiency Testing Samples from RTC


PT Samples from RTCRTC has been producing environmental LPTP for more than 20 years and send out over 20,000 PT samples a year, throughout
the world.





Customers Also Viewed

Benzenethiol

≥98%

Thiophenol

purum, ≥98.0% (GC)

Thiophenol

≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?