• USA Home
  • 09820 - Ammonium hexafluorophosphate

EMAIL THIS PAGE TO A FRIEND
09820 Fluka

Ammonium hexafluorophosphate

≥98.0% (as NH4PF6, gravimetric)

Purchase

Properties

Related Categories Ammonium, Ammonium Salts, Chemical Synthesis, Essential Chemicals, Inorganic Salts,
assay   ≥98.0% (as NH4PF6, gravimetric)
solubility   H2O: soluble50 mg/mL, clear, colorless
density   2.18 g/mL at 25 °C(lit.)
Featured Industry   Battery Manufacturing

Description

Legal Information

FLUKA is a registered trademark of Honeywell Specialty Chemicals Seelze GmbH.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3260 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?