CAS Number 63148-64-1
MDL number MFCD00131103
| Related Categories | Analytical Reagents, Analytical/Chromatography, Fixation Reagents, Fixatives, Hematology and Histology, |
| grade | for microscopy |
| concentration | ~1% in 1,2-dichloroethane |
| refractive index |
n |
| density | 1.253 g/mL at 20 °C |
Customers Also Viewed
~0.25% in 1,2-dichloroethane, for microscopy
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?