CAS Number 51636-39-6
MDL number MFCD00676481
PubChem Substance ID 24846996
| Related Categories | Analytical Reagents, Analytical/Chromatography, Application Specific Reagents, Decomposition Reagents, Kjeldahl Catalysts & Oxidants for Digestion, |
| composition | Cu, ~60% |
| Mg, ~40% |
reducing agent for the determination of nitrogen in the neutral to weak acid pH-range
Alloy (60% Cu, 40% Mg) for the reduction of nitrites and nitrates in weakly acidic or neutral solution for analysis (alternative to Devarda alloy)1
Customers Also Viewed
for determination of nitrogen
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?